5-Isopropyl[1,3,4]thiadiazol-2-ylamine 96%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-isopropyl-1,3,4-thiadiazole

A654601-1G A654601-250MG
76775-578EA 45.62 USD
76775-578 76775-580
5-Isopropyl[1,3,4]thiadiazol-2-ylamine 96%
5-Isopropyl[1,3,4]thiadiazol-2-ylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃S
MW: 143.21 g/mol
MDL Number: MFCD00466388
CAS Number: 27115-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR