4-tert-Butylcalix[6]arene 98%

Supplier: Ambeed
Warning

A649906-1G A649906-25G A649906-5G
76775-554EA 29 USD
76775-554 77598-652 77598-654
4-tert-Butylcalix[6]arene 98%
4-tert-Butylcalix[6]arene

Precaución: for Research Use Only

Formula: C₆₆H₈₄O₆
MDL Number: MFCD00075464
CAS Number: 78092-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR