6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A648543-1G A648543-5G
76775-550EA 44 USD
76775-550 76775-552
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine 95%
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD00763284
CAS Number: 42869-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR