trans,trans-2,4-Heptadienal 90%

Supplier: Ambeed
Danger

Synonyms: (2E,4E)-Hepta-2,4-dienal

A645328-10G A645328-1G A645328-25G A645328-5G
76775-530EA 34 USD
76775-530 76775-532 76775-534 76775-536
trans,trans-2,4-Heptadienal 90%
trans,trans-2,4-Heptadienal

Precaución: for Research Use Only

Formula: C₇H₁₀O
MW: 110.16 g/mol
MDL Number: MFCD00007005
CAS Number: 4313-03-5
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR