Methyl-3-amino-5-phenylthiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A644749-100G A644749-25G A644749-5G
76775-520EA 322 USD
76775-520 76775-522 76775-524
Methyl-3-amino-5-phenylthiophene-2-carboxylate 98%
Methyl-3-amino-5-phenylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂S
MDL Number: MFCD00068161
CAS Number: 100063-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR