Cyclopropyl phenyl ketone 95%

Supplier: Ambeed
Warning

A642851-100G A642851-1G A642851-25G A642851-500G A642851-5G
76775-510EA 208 USD
76775-510 76775-512 76775-514 76775-516 76775-518
Cyclopropyl phenyl ketone 95%
Cyclopropyl phenyl ketone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
MDL Number: MFCD00001294
CAS Number: 3481-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR