6-Methyl-5-nitro-1H-indazole 96%

Supplier: Ambeed
Warning

A641645-100MG A641645-1G A641645-250MG
76775-504EA 21.22 USD
76775-504 76775-506 76775-508
6-Methyl-5-nitro-1H-indazole 96%
6-Methyl-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.1601 g/mol
MDL Number: MFCD03305368
CAS Number: 81115-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR