5-Methyl-2-furonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 5-Methylfuran-2-carbonitrile

A633925-100MG A633925-1G A633925-250MG A633925-5G
76775-470EA 12.74 USD
76775-470 76775-472 76775-474 76775-476
5-Methyl-2-furonitrile 95%
5-Methyl-2-furonitrile

Precaución: for Research Use Only

Formula: C₆H₅NO
MW: 107.11 g/mol
MDL Number: MFCD02093782
CAS Number: 13714-86-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR