6-Bromo-2,2'-bipyridyl 97%

Supplier: Ambeed
Danger

A629760-100MG A629760-1G A629760-250MG A629760-5G A629760-25G
76775-456EA 19 USD
76775-456 76775-458 76775-460 76775-462 77569-202
6-Bromo-2,2'-bipyridyl 97%
6-Bromo-2,2'-bipyridyl

Precaución: For research use only.

Formula: C₁₀H₇BrN₂
MDL Number: MFCD01318953
CAS Number: 10495-73-5
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR