4'-(Diethylamino)acetophenone 98%

Supplier: Ambeed
Warning

A628642-5G A628642-1G
76775-454EA 104 USD
76775-454 77598-618
4'-(Diethylamino)acetophenone 98%
4'-(Diethylamino)acetophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
MDL Number: MFCD00043651
CAS Number: 5520-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR