4-tert-Butylcalix[4]arene 98%

Supplier: Ambeed
Warning

A626187-1G A626187-5G A626187-100G
76775-444EA 9 USD
76775-444 77551-876 77627-258
4-tert-Butylcalix[4]arene 98%
4-tert-Butylcalix[4]arene

Precaución: for Research Use Only

Formula: C₄₄H₅₆O₄
MDL Number: MFCD00066280
CAS Number: 60705-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR