Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
A624979-1G
A624979-250MG
A624979-25G
A624979-5G
76775-424EA
15.92
USD
76775-424
76775-426
76775-428
76775-430
Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₂FN₃O₂ MW: 249.2410 g/mol |
MDL Number:
MFCD00173917 CAS Number: 138907-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...