6-Methylbenzo[d]oxazole 98%

Supplier: Ambeed
Warning

A624502-1G A624502-250MG A624502-5G
76775-418EA 28 USD
76775-418 76775-420 76775-422
6-Methylbenzo[d]oxazole 98%
6-Methylbenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₇NO
MDL Number: MFCD00216937
CAS Number: 10531-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR