N-(Hydroxymethyl)nicotinamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(Hydroxymethyl)-3-pyridinecarboxamide
A623137-100MG
A623137-1G
76775-392EA
5.31
USD
76775-392
76775-394
N-(Hydroxymethyl)nicotinamide 97%
N-(Hydroxymethyl)nicotinamide
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol |
MDL Number:
MFCD00010437 CAS Number: 3569-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...