5-Chloro-7-nitro-1H-indazole 97%

Supplier: Ambeed
Danger

A622300-1G A622300-250MG
76775-384EA 172.93 USD
76775-384 76775-386
5-Chloro-7-nitro-1H-indazole 97%
5-Chloro-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.57 g/mol
MDL Number: MFCD09027005
CAS Number: 41926-18-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR