3-Bromo-5-methylbenzeneboronic acid 97%

Supplier: Ambeed
Warning

A621328-10G A621328-25G A621328-5G A621328-1G
76775-378EA 60 USD
76775-378 76775-380 76775-382 77878-435
3-Bromo-5-methylbenzeneboronic acid 97%
3-Bromo-5-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MDL Number: MFCD06411355
CAS Number: 849062-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR