Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

A620725-10G A620725-1G A620725-25G A620725-5G
76775-370EA 91.24 USD
76775-370 76775-372 76775-374 76775-376
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate 97%
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂S
MW: 240.33 g/mol
MDL Number: MFCD01147322
CAS Number: 24237-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR