2-(Aminomethyl)-1-ethylpyrrolidine 98%

Supplier: Ambeed
Danger

A619028-10G A619028-25G A619028-5G
76775-358EA 8.49 USD
76775-358 76775-360 76775-362
2-(Aminomethyl)-1-ethylpyrrolidine 98%
2-(Aminomethyl)-1-ethylpyrrolidine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD00003178
CAS Number: 26116-12-1
UN: 2733
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR