5-(4-Methylphenyl)cyclohexane-1,3-dione 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(p-Tolyl)cyclohexane-1,3-dione
A617938-100MG
A617938-1G
A617938-250MG
76775-348EA
31.83
USD
76775-348
76775-350
76775-352
5-(4-Methylphenyl)cyclohexane-1,3-dione 97%
5-(4-Methylphenyl)cyclohexane-1,3-dione
Precaución: for Research Use Only
Formula:
C₁₃H₁₄O₂ MW: 202.26 g/mol |
MDL Number:
MFCD00111717 CAS Number: 61888-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...