6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-8-methyl-[1,2,4]triazolo[1,5-a]pyridine

A612285-1G A612285-5G
76775-308EA 227.04 USD
76775-308 76775-310
6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine 95%
6-Bromo-8-methyl[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD09038166
CAS Number: 899429-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR