4-Amino-3-(trifluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A610280-10G A610280-1G A610280-250MG A610280-25G A610280-5G
76775-276EA 77.45 USD
76775-276 76775-278 76775-280 76775-282 76775-284
4-Amino-3-(trifluoromethoxy)benzoic acid 97%
4-Amino-3-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MDL Number: MFCD00205143
CAS Number: 175278-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR