1-Boc-(2-aminomethyl)piperidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl Ester, 2-Aminomethylpiperidine-1-carboxylic acid tert-butyl ester, 1-Boc-2-aminomethylpiperidine, 2-(Aminomethyl)-1-Boc-piperidine, 2-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 1-Boc-2-(Aminomethyl)piperidine, 1,1-Dimethylethyl 2-(aminomethyl)-1-piperidinecarboxylate, 2-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 2-(Aminomethyl)-1-piperidinecarboxylate, 2-Aminomethyl-1-Boc-piperidine

A610203-10G A610203-1G A610203-250MG A610203-5G
76775-268EA 136 USD
76775-268 76775-270 76775-272 76775-274
1-Boc-(2-aminomethyl)piperidine 98%
1-Boc-(2-aminomethyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD01317791
CAS Number: 370069-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR