5-Bromo-7-chloroindole 96%

Supplier: Ambeed
Warning

A608816-250MG A608816-1G A608816-5G
76775-266EA 64.72 USD
76775-266 77572-982 77594-862
5-Bromo-7-chloroindole 96%
5-Bromo-7-chloroindole

Precaución: for Research Use Only

Formula: C₈H₅BrClN
MW: 230.48 g/mol
MDL Number: MFCD03094978
CAS Number: 180623-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR