5-Methyl-1H-pyrrole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methylpyrrole-2-carbaldehyde

A608700-1G A608700-250MG A608700-5G
76775-260EA 110.34 USD
76775-260 76775-262 76775-264
5-Methyl-1H-pyrrole-2-carbaldehyde 95%
5-Methyl-1H-pyrrole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.126 g/mol
MDL Number: MFCD08236773
CAS Number: 1192-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR