Z-Pro-ONp 95+%

Supplier: Ambeed
Warning

A608513-10G A608513-1G A608513-5G
76775-254EA 166.57 USD
76775-254 76775-256 76775-258
Z-Pro-ONp 95+%
Z-Pro-ONp

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₆
MDL Number: MFCD00020828
CAS Number: 3304-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR