5-Bromo-α,α,α-trifluoro-2-nitrotoluene 98%

Supplier: Ambeed
Warning

A607342-100G A607342-10G A607342-1G A607342-25G A607342-5G
76775-244EA 36 USD
76775-244 76775-246 76775-248 76775-250 76775-252
5-Bromo-α,α,α-trifluoro-2-nitrotoluene 98%
5-Bromo-α,α,α-trifluoro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD00014706
CAS Number: 344-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR