Benzyl-5-bromoamyl ether 95%

Supplier: Ambeed
Warning

Synonyms: (((5-Bromopentyl)oxy)methyl)benzene, (5-Bromopentyloxymethyl)benzene, Benzyloxypentyl Bromide, Benzyl 5-Bromopentyl Ether

A605503-1G A605503-25G A605503-5G
76775-228EA 15 USD
76775-228 76775-230 77569-152
Benzyl-5-bromoamyl ether 95%
Benzyl-5-bromoamyl ether

Precaución: For research use only.

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
MDL Number: MFCD02258456
CAS Number: 1014-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR