2-Bromo-4,6-xylidine 98%

Supplier: Ambeed
Warning

A604389-1G A604389-25G A604389-5G
76775-216EA 5 USD
76775-216 76775-218 76775-220
2-Bromo-4,6-xylidine 98%
2-Bromo-4,6-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MDL Number: MFCD00047826
CAS Number: 41825-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR