8-Nitroquinaldine 98%

Supplier: Ambeed
Danger

A604055-10G A604055-1G A604055-25G A604055-5G
76775-210EA 46 USD
76775-210 76775-212 76775-214 77878-403
8-Nitroquinaldine 98%
8-Nitroquinaldine

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00006764
CAS Number: 881-07-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR