6-Fluoro-1H-indazole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A602746-100MG A602746-1G A602746-250MG
76775-198EA 125.19 USD
76775-198 76775-200 76775-202
6-Fluoro-1H-indazole-3-carbaldehyde 98%
6-Fluoro-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MW: 164.13 g/mol
MDL Number: MFCD07781606
CAS Number: 518987-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR