2-Fluoro-5-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A602300-1G A602300-25G A602300-5G A602300-100G
76775-192EA 6 USD
76775-192 76775-194 76775-196 77878-399
2-Fluoro-5-iodobenzonitrile 98%
2-Fluoro-5-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
MDL Number: MFCD03094225
CAS Number: 351003-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR