2,4,5-Trifluorobenzaldehyde 98%

Supplier: Ambeed
Danger

A600064-100G A600064-10G A600064-1G A600064-25G A600064-5G
76775-182EA 117 USD
76775-182 76775-184 76775-186 76775-188 76775-190
2,4,5-Trifluorobenzaldehyde 98%
2,4,5-Trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₃O
MDL Number: MFCD00061196
CAS Number: 165047-24-5
UN: 1989
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR