5-Aminopentan-1-ol 95%

Supplier: Ambeed
Danger

Synonyms: 5-Amino-1-pentanol

A597839-10G A597839-25G A597839-5G A597839-100G
76775-172EA 20.16 USD
76775-172 76775-174 76775-176 77627-232
5-Aminopentan-1-ol 95%
5-Aminopentan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00008237
CAS Number: 2508-29-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR