(±)-δ-Nonalactone 95%

Supplier: Ambeed
Danger

A596256-5G A596256-25G
76775-156EA 8 USD
76775-156 77878-394
(±)-δ-Nonalactone 95%
(±)-δ-Nonalactone

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MDL Number: MFCD00036500
CAS Number: 3301-94-8
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR