Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate 98%

Supplier: Ambeed
Danger

A594574-25G A594574-5G
76775-146EA 24 USD
76775-146 77878-393
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate 98%
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00274087
CAS Number: 96562-58-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR