2,6-Difluorobenzylamine 98%

Supplier: Ambeed
Danger

A592624-10G A592624-1G A592624-25G A592624-5G
76775-138EA 26.53 USD
76775-138 76775-140 76775-142 76775-144
2,6-Difluorobenzylamine 98%
2,6-Difluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.14 g/mol
MDL Number: MFCD00010144
CAS Number: 69385-30-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR