5-Chloropent-1-ene 97%

Supplier: Ambeed
Danger

A592079-10G A592079-1G A592079-25G A592079-5G
76775-130EA 41.38 USD
76775-130 76775-132 76775-134 76775-136
5-Chloropent-1-ene 97%
5-Chloropent-1-ene

Precaución: for Research Use Only

Formula: C₅H₉Cl
MW: 104.57 g/mol
MDL Number: MFCD00039389
CAS Number: 928-50-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR