(±)-γ-Dodecalactone 98%

Supplier: Ambeed
Warning

A589642-100G A589642-25G A589642-500G A589642-5G
76775-122EA 44 USD
76775-122 76775-124 76775-126 76775-128
(±)-γ-Dodecalactone 98%
(±)-γ-Dodecalactone

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00036499
CAS Number: 2305-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR