6-Chloro-1,2-benzisoxazol-3-ol 95%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-hydroxy-1,2-benzisoxazole

A588293-100MG A588293-1G A588293-250MG A588293-5G
76775-092EA 33.95 USD
76775-092 76775-094 76775-096 76775-098
6-Chloro-1,2-benzisoxazol-3-ol 95%
6-Chloro-1,2-benzisoxazol-3-ol

Precaución: for Research Use Only

Formula: C₇H₄ClNO₂
MW: 169.57 g/mol
MDL Number: MFCD00125033
CAS Number: 61977-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR