Inosine-5'-monophosphate Disodium Salt octahydrate 98%
Supplier: Ambeed
|
Warning
|
A587717-100G
A587717-1G
A587717-25G
A587717-5G
76775-084EA
59.42
USD
76775-084
76775-086
76775-088
76775-090
Inosine-5'-monophosphate Disodium Salt octahydrate 98%
Inosine-5'-monophosphate Disodium Salt octahydrate
Precaución: for Research Use Only
Formula:
C₁₀H₂₇N₄Na₂O₁₆P MW: 536.29 g/mol |
MDL Number:
MFCD29913091 CAS Number: 20813-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...