4-[(2-Hydroxyethyl)amino]-3-nitrophenol 98%

Supplier: Ambeed
Warning

A587226-100G A587226-25G A587226-500G A587226-5G
76775-076EA 101 USD
76775-076 76775-078 76775-080 76775-082
4-[(2-Hydroxyethyl)amino]-3-nitrophenol 98%
4-[(2-Hydroxyethyl)amino]-3-nitrophenol

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₄
MDL Number: MFCD02684022
CAS Number: 65235-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR