4-Amino-3-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-2-nitroaniline

A584690-100G A584690-1G A584690-25G A584690-5G A584690-500G
76775-064EA 221 USD
76775-064 76775-066 76775-068 76775-070 76775-072 77878-385
4-Amino-3-nitrobenzonitrile 97%
4-Amino-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD00013373
CAS Number: 6393-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR