α,α,α-Trifluoro-3,5-diaminotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 5-(Trifluoromethyl)-1,3-phenylenediamine, 5-(Trifluoromethyl)-m-phenylenediamine, 5-Trifluoromethyl-1,3-phenylenediamine, 5-(Trifluoromethyl)benzene-1,3-diamine, 3,5-Diaminobenzotrifluoride

A583810-1G A583810-25G
76775-060EA 15 USD
76775-060 76775-062
α,α,α-Trifluoro-3,5-diaminotoluene 98%
α,α,α-Trifluoro-3,5-diaminotoluene

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂
MDL Number: MFCD00014776
CAS Number: 368-53-6
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR