4-Bromo-2-(difluoromethyl)-1-fluorobenzene 98%

Supplier: Ambeed
Warning

A578741-5G A578741-25G A578741-1G
76775-046EA 53 USD
76775-046 77569-104 77878-377
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 98%
4-Bromo-2-(difluoromethyl)-1-fluorobenzene

Precaución: For research use only.

Formula: C₇H₄BrF₃
MDL Number: MFCD06411633
CAS Number: 445303-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR