Isoamyl valerate 98%

Supplier: Ambeed
Warning

Synonyms: Isopentyl pentanoate

A577946-100G A577946-25G A577946-500G
76775-034EA 10.61 USD
76775-034 76775-036 76775-038
Isoamyl valerate 98%
Isoamyl valerate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD00085198
CAS Number: 2050-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR