2-Bromo-6-ethoxynaphthalene 98%

Supplier: Ambeed
Warning

A570746-1G A570746-250MG A570746-5G
76775-004EA 31.83 USD
76775-004 76775-006 76775-008
2-Bromo-6-ethoxynaphthalene 98%
2-Bromo-6-ethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrO
MDL Number: MFCD01568865
CAS Number: 66217-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR