3-Fluoro-o-toluonitrile 98%

Supplier: Ambeed
Danger

A570612-1G A570612-5G
76775-000EA 6 USD
76775-000 76775-002
3-Fluoro-o-toluonitrile 98%
3-Fluoro-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MDL Number: MFCD03788541
CAS Number: 185147-06-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR