Ethyl 5,7-dichloro-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A568912-10G A568912-1G A568912-250MG A568912-5G
76774-992EA 237.65 USD
76774-992 76774-994 76774-996 76774-998
Ethyl 5,7-dichloro-1H-indole-2-carboxylate 97%
Ethyl 5,7-dichloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉Cl₂NO₂
MW: 258.10 g/mol
MDL Number: MFCD03084730
CAS Number: 4792-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR