6-Methoxybenzo[d]thiazole-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxybenzothiazole-2-carboxamide

A567936-1G A567936-5G
76774-990EA 21.22 USD
76774-990 77627-172
6-Methoxybenzo[d]thiazole-2-carboxamide 95%
6-Methoxybenzo[d]thiazole-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MW: 208.237 g/mol
MDL Number: MFCD00297288
CAS Number: 946-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR