6-Methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A565906-1G A565906-250MG
76774-988EA 268.41 USD
76774-988 77627-166
6-Methylbenzo[d]thiazole 95%
6-Methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₇NS
MW: 149.21 g/mol
MDL Number: MFCD01325556
CAS Number: 2942-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR